(3,4-Dimethoxyphenyl)acetonitrile 99+%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

155260250 155261000 155265000
ACRO155260250EA 18 GBP
ACRO155260250 ACRO155261000 ACRO155265000
(3,4-Dimethoxyphenyl)acetonitrile 99+%
(3,4-Dimethoxyphenyl)acetonitrile
Formula: C₁₀H₁₁NO₂
Boiling Pt: 171…178 °C (10 mmHg)
Melting Pt: 62…65 °C
Flash Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00001911
CAS Number: 93-17-4
EINECS: 202-225-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to off-white Crystals or crystalline needles
Infrared spectrum Conforms
Melting point 62 to 65 °C
Loss on drying ≤0.5 % (vacuum)
Sulfated ash ≤0.1 %
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR