Cyclopropylamine 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

154880100 154880500 154882500
ACRO154880100EA 27.7 GBP
ACRO154880100 ACRO154880500 ACRO154882500
Cyclopropylamine 99%
Cyclopropylamine
Formula: C₃H₇N
Boiling Pt: 49…50 °C (760 mmHg)
Melting Pt: –50 °C
Density: 0.824 g/cm³
Flash Pt: –25 °C
Storage Temperature: Ambient
MDL Number: MFCD00001301
CAS Number: 765-30-0
EINECS: 212-142-2
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Volatile liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4220 (20°C, 589 nm)
Water ≤0.5 %
GC ≥99 %
Titration with HCl 99.0 to 101.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR