Phenylacetylene 98%, pure

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

152460250 152461000 152465000
ACRO152460250EA 28 GBP
ACRO152460250 ACRO152461000 ACRO152465000
Phenylacetylene 98%, pure
Phenylacetylene
Formula: C₈H₆
MW: 102.14 g/mol
Boiling Pt: 142…144 °C (760 mmHg)
Melting Pt: –44.8 °C
Density: 0.93 g/cm³
Flash Pt: 31 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008570
CAS Number: 536-74-3
EINECS: 208-645-1
UN: 3295
ADR: 3 III
Merck Index: 13,03892

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5470 to 1.5500 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR