Amantadine hydrochloride 99+%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

151670250 151671000
ACRO151670250EA 33.9 GBP
ACRO151670250 ACRO151671000
Amantadine hydrochloride 99+%
Amantadine hydrochloride
Formula: C₁₀H₁₈ClN
Melting Pt: >300 °C
MDL Number: MFCD00074723
CAS Number: 665-66-7
EINECS: 211-560-2

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥99.0 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR