Methyl yellow, pure indicator grade

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Dimethyl yellow, Sudan Yellow, 4-(Dimethylamino)azobenzene, 4-(N,N-Dimethylamino)azobenzene

151401000 151400250 151405000
ACRO151401000EA 42.3 GBP
ACRO151401000 ACRO151400250 ACRO151405000
Methyl yellow, pure indicator grade
Methyl Yellow
Formula: C₁₄H₁₅N₃
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 111…117 °C
Density: 1,2233 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008308
CAS Number: 60-11-7
EINECS: 200-455-7
UN: 3143
ADR: 6.1,III
Merck Index: 12,03279

Order Now

Specification Test Results

Appearance Orange to orange-brown Powder
Infrared spectrum Conforms
Loss on drying ≤3 %
Appearance of solution (0.1% soln. in ethanol) clear
Visual transition interval from pH 2.9 (red) to pH 4.0 (yellow)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR