Benzocaine (ethyl-4-aminobenzoate) 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

150781000 150785000 150780025
ACRO150781000EA 34.7 GBP
ACRO150781000 ACRO150785000 ACRO150780025
Benzocaine (ethyl-4-aminobenzoate) 98%
Benzocaine (Ethyl 4-aminobenzoate)
Formula: C₉H₁₁NO₂
Boiling Pt: 172 °C (17hPa)
Melting Pt: 89…92 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00007892
CAS Number: 94-09-7
EINECS: 202-303-5
Merck Index: 13,01087

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 89 to 92 °C
GC ≥97.5 %
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR