1,1,1-Trichloro-2-methyl-2-propanol hemihydrate 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 1,1,1-Trichloro-2-methyl-2-propanol Hemihydrate, Chlorbutanol hemihydrate, Chlorobutanol hemihydrate, Chlorobutanol hydrous

150305000 150301000 150300025
ACRO150305000EA 67.7 GBP
ACRO150305000 ACRO150301000 ACRO150300025
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate 98%
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate
Formula: C₈H₁₆Cl₆O₃
Boiling Pt: 173…175 °C (760 mmHg)
Melting Pt: 75…79 °C
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD02179352
CAS Number: 6001-64-5
EINECS: 200-317-6

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration Argentometric 97.5 to 102.5 % (ex Cl)
Water 4.5 to 5.5 % (K.F.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR