3-Hydroxypropiononitrile 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

149390010 149390500 149392500
ACRO149390010EA 221 GBP
ACRO149390010 ACRO149390500 ACRO149392500
3-Hydroxypropiononitrile 99%
3-Hydroxypropiononitrile
Formula: C₃H₅NO
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: –46 °C
Density: 1.04 g/cm³
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00002826
CAS Number: 109-78-4
EINECS: 203-704-8
Merck Index: 13,03828

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4246 to 1.4266 (20°C, 589 nm)
GC ≥98.5 %
Specific optical rotation -28° to -32° (20°C, 589 nm) (c=1 in DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR