(±)-4-Methyl-2-pentanol 99+%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

149380010 149380025 149380050 149385000 149380100
ACRO149380010EA 30.1 GBP
ACRO149380010 ACRO149380025 ACRO149380050 ACRO149385000 ACRO149380100
(±)-4-Methyl-2-pentanol 99+%
(±)-4-Methyl-2-pentanol
Formula: C₆H₁₄O
Boiling Pt: 132 °C (760 mmHg)
Melting Pt: –90 °C
Density: 0.8 g/cm³
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00004550
CAS Number: 108-11-2
EINECS: 203-551-7
UN: 2053
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4100 to 1.4120 (20°C, 589 nm)
GC ≥99.0 %
Specific optical rotation +28° to +32° (20°C, 589 nm) (c=1 in DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR