Diethylene glycol monomethyl ether 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2-(2-Methoxyethoxy)ethanol, Diethylene glycol methyl ether

149370010 149370025 149370100
ACRO149370010EA 29.7 GBP
ACRO149370010 ACRO149370025 ACRO149370100
Diethylene glycol monomethyl ether 99%
Diethylene glycol monomethyl ether
Formula: C₅H₁₂O₃
Boiling Pt: 194 °C (760 mmHg)
Melting Pt: –70 °C
Density: 1.01 g/cm³
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00002871
CAS Number: 111-77-3
EINECS: 203-906-6
Merck Index: 12,06116

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4250 to 1.4270 (20°C, 589 nm)
Water ≤0.2 %
GC ≥98.5 %
Color scale ≤15 APHA

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR