Methyl phenylacetate 99+%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

148440050 148442500 148440010
ACRO148440050EA 23.2 GBP
ACRO148440050 ACRO148442500 ACRO148440010
Methyl phenylacetate 99+%
Methyl phenylacetate
Formula: C₉H₁₀O₂
Boiling Pt: 218 °C (1013 hPa)
Melting Pt: 49…50 °C
Density: 1.067 g/cm³
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00008453
CAS Number: 101-41-7
EINECS: 202-940-9
Merck Index: 12,07422

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5060 to 1.5080 (20°C, 589 nm)
GC ≥99.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR