4-(Dimethylamino)pyridine 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 4-Dimethylaminopyridine, DMAP, N,N-Dimethylpyridin-4-amine

148270050 148270250 148271000 148270025 148275000
ACRO148270050EA 14.9 GBP
ACRO148270050 ACRO148270250 ACRO148271000 ACRO148270025 ACRO148275000
4-(Dimethylamino)pyridine 99%
4-(Dimethylamino)pyridine
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 162 °C (50 mmHg)
Melting Pt: 110…113 °C
Flash Pt: 124 °C
Storage Temperature: Ambient
MDL Number: MFCD00006418
CAS Number: 1122-58-3
EINECS: 214-353-5
UN: 2811
ADR: 6.1 II

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 110 to 113 °C
Water ≤0.1 %
GC ≥98.5 %
Color of alcohol solution ≤30 APHA (10% W/V in ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR