3-Amino-9-ethyl carbazole (AEC) 90% tech.

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

147870050 147870250 147871000
ACRO147870050EA 21.5 GBP
ACRO147870050 ACRO147870250 ACRO147871000
3-Amino-9-ethyl carbazole (AEC) 90% tech.
3-Amino-9-ethyl carbazole
Formula: C₁₄H₁₄N₂
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004964
CAS Number: 132-32-1
EINECS: 205-057-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Brown Crystalline powder and/or chunks
Infrared spectrum Conforms
GC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR