3-Bromothiophene 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

147800250 147801000 147805000
ACRO147800250EA 46.5 GBP
ACRO147800250 ACRO147801000 ACRO147805000
3-Bromothiophene 97%
3-Bromothiophene
Formula: C₄H₃BrS
Boiling Pt: 158 °C (760 mmHg)
Melting Pt: -10 °C
Density: 1.74 g/cm³
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00005464
CAS Number: 872-31-1
EINECS: 212-821-3
UN: 3384
ADR: 6.1,

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5905 to 1.5925 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR