Adenine 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Purin-6-amine, 7H-Purin-6-amine, 6-Aminopurine

147440250 147441000 147445000
ACRO147440250EA 85.7 GBP
ACRO147440250 ACRO147441000 ACRO147445000
Adenine 99%
Adenine
Formula: C₅H₅N₅
Melting Pt: 360…365 °C (dec.)
Density: 0,99741 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041790
CAS Number: 73-24-5
EINECS: 200-796-1
UN: 2811
ADR: 6.1,III
Merck Index: 13,00152

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C, 5 hrs)
Sulfated ash ≤0.1 %
HPLC ≥98.5 %
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR