2,4-Diaminotoluene 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

146600010 146600250 146602500
ACRO146600010EA 493 GBP
ACRO146600010 ACRO146600250 ACRO146602500
2,4-Diaminotoluene 98%
2,4-Diaminotoluene
Formula: C₇H₁₀N₂
Boiling Pt: 283…285 °C (760 mmHg)
Melting Pt: 97…101 °C
Density: 1,045 g/cm³ (20 °C)
Flash Pt: 149 °C
Storage Temperature: Ambient
MDL Number: MFCD00007804
CAS Number: 95-80-7
EINECS: 202-453-1
UN: 1709
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Dark grey to brown Powder or crystals or flakes and/or chunks
Infrared spectrum Conforms
Melting point 95 to 102 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR