Thioanisole 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Methyl phenyl sulphide, Phenyl methyl sulphide, (Methylthio)benzene

138670250 138671000 138675000 138670025
ACRO138670250EA 23.8 GBP
ACRO138670250 ACRO138671000 ACRO138675000 ACRO138670025
Thioanisole 99%
Thioanisole
Formula: C₇H₈S
Boiling Pt: 188 °C (760 mmHg)
Melting Pt: –15 °C
Density: 1.05 g/cm³
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00008559
CAS Number: 100-68-5
EINECS: 202-878-2
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5850 to 1.5880 (20°C, 589 nm)
GC ≥98.5 %
Specific optical rotation -8° to -12° (20°C, 589 nm) (c=1, chloroform)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR