Charcoal activated, NORIT® SA 2

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Carbon activated, Activated carbon, Activated charcoal

134370025 134372500 134370250
ACRO134370025EA 116 GBP
ACRO134370025 ACRO134372500 ACRO134370250
Charcoal activated, NORIT® SA 2
Charcoal activated
Formula: C
MW: 12.01 g/mol
Melting Pt: 3652 °C
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
Merck Index: 13,01818

Order Now

Specification Test Results

Appearance Black Powder
Loss on drying ≤10 %
Molasses number ≤390
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR