Nicotinic acid (Niacin) 99.5%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

128290050 128291000 128295000 128290025 128290100
ACRO128290050EA 22.6 GBP
ACRO128290050 ACRO128291000 ACRO128295000 ACRO128290025 ACRO128290100
Nicotinic acid (Niacin) 99.5%
Nicotinic acid (Niacin)
Formula: C₆H₅NO₂
Boiling Pt: 258 °C (1013 hPa)
Melting Pt: 236…239 °C
Density: 1,473 g/cm³ (20 °C)
Flash Pt: 193 °C
Storage Temperature: Ambient
MDL Number: MFCD00006391
CAS Number: 59-67-6
EINECS: 200-441-0
Merck Index: 13,06552

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 234 to 240 °C
Loss on drying ≤1 % (105 °C, 1 h)
Sulfated ash ≤0.1 %
Titration with NaOH ≥99.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR