Nickel(II) acetylacetonate 96%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

128260250 128260050 128261000
ACRO128260250EA 61.6 GBP
ACRO128260250 ACRO128260050 ACRO128261000
Nickel(II) acetylacetonate 96%
Nickel (II) acetylacetonate
Formula: C₁₀H₁₄NiO₄
Boiling Pt: 220 °C (11 mmHg)
Melting Pt: 230 °C (dec.)
Density: 1,455 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000024
CAS Number: 3264-82-2
EINECS: 221-875-7
Merck Index: 12,06584

Order Now

Specification Test Results

Appearance Green Powder
Infrared spectrum Conforms
Titration Complexometric ≥95 %
Chloride (Cl) ≤5 ppm
Phosphate (PO4) ≤5 ppm
Calcium (Ca) ≤10 ppm
Copper (Cu) ≤5 ppm
Iron (Fe) ≤5 ppm
Potassium (K) ≤10 ppm
Sodium (Na) ≤20 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 % (in water)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR