Menthone (mixture of isomers) 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Menthone, 2-Isopropyl-5-Methylcyclohexanone

125411000
ACRO125411000EA 86.1 GBP
ACRO125411000
Menthone (mixture of isomers) 98%
Menthone (mixture of isomers)
Formula: C₁₀H₁₈O
Melting Pt: –6 °C
Density: 0.89 g/cm³
Flash Pt: 69 °C
Storage Temperature: Ambient
MDL Number: MFCD00062998
CAS Number: 10458-14-7
EINECS: 233-944-9

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4460 to 1.4550 (20°C, 589 nm)
Density (20°C) 0.889 to 0.898 g/ml
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR