Oxybenzone 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

121360050 121361000
ACRO121360050EA 14.6 GBP
ACRO121360050 ACRO121361000
Oxybenzone 98%
Oxybenzone
Formula: C₁₄H₁₂O₃
Boiling Pt: 150…160 °C (5 mmHg)
Melting Pt: >62.5 °C
Density: 1,3397 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00008387
CAS Number: 131-57-7
EINECS: 205-031-5
Merck Index: 12,07088

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Melting point ≥62.0 °C
GC ≥97.5 %
Specific extinction E (1%/1cm) ≥400 (321 to 327 nm, methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR