Caproic acid 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

120700010 120705000 120700025 120702500
ACRO120700010EA 61.4 GBP
ACRO120700010 ACRO120705000 ACRO120700025 ACRO120702500
Caproic acid 99%
Caproic acid
Formula: C₆H₁₂O₂
Boiling Pt: 202…203 °C (760 mmHg)
Melting Pt: –3 °C
Density: 0.92 g/cm³
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00004421
CAS Number: 142-62-1
EINECS: 205-550-7
UN: 2829
ADR: 8,III
Merck Index: 13,01765

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4150 to 1.4180 (20°C, 589 nm)
GC ≥98.5 %
Acid value 473 to 493 mg KOH/g
test 11 Result 11
test 12 Result 12
test 13 Result 13
test 14 Result 14
test 15 Result 15
test 16 Result 16
test 17 Result 17
test 18 Result 18
test 19 Result 19
test 20 Result 20
test 21 Result 21
test 22 Result 22
test 23 Result 23
test 24 Result 24
test 25 Result 25
test 26 Result 26
test 27 Result 27
test 28 Result 28
test 29 Result 29
test 30 Result 30
test 31 Result 31
test 32 Result 32
test 33 Result 33
test 34 Result 34
test 35 Result 35
test 36 Result 36
test 37 Result 37
test 38 Result 38
test 39 Result 39
test 40 Result 40
test 41 Result 41
test 42 Result 42
test 43 Result 43
test 44 Result 44
test 45 Result 45
test 46 Result 46
test 47 Result 47
test 48 Result 48
test 49 Result 49
test 50 Result 50
test 51 Result 51
test 52 Result 52
test 53 Result 53
test 54 Result 54
test 55 Result 55
test 56 Result 56
test 57 Result 57


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR