1-Hexadecanol 96%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Cetyl alcohol, Palmityl alcohol

120480010 120480025 120480100
ACRO120480010EA 42.9 GBP
ACRO120480010 ACRO120480025 ACRO120480100
1-Hexadecanol 96%
1-Hexadecanol
Formula: H₃C(CH₂)₁₄CH₂OH
MW: 242.45 g/mol
Boiling Pt: 344 °C (760 mmHg)
Melting Pt: 47…50 °C
Flash Pt: >157 °C
Storage Temperature: Ambient
MDL Number: MFCD00004760
CAS Number: 36653-82-4
EINECS: 253-149-0
Merck Index: 13,02037

Order Now

Specification Test Results

Appearance White to off-white Powder or flakes or pellets
Infrared spectrum Conforms
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR