L(-)-Glutathione (reduced form) 98% for analysis, reduced

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

120000010 120000050 120000250 120001000 120005000
ACRO120000010EA 26.2 GBP
ACRO120000010 ACRO120000050 ACRO120000250 ACRO120001000 ACRO120005000
L(-)-Glutathione (reduced form) 98% for analysis, reduced
L(-)-Glutathione (reduced form)
Formula: C₁₀H₁₇N₃O₆S
Melting Pt: 182…192 °C (dec.)
Density: 1,475 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00065939
CAS Number: 70-18-8
EINECS: 200-725-4
Merck Index: 13,04488

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 182 to 192 °C (Dec)
Titration Iodimetric ≥97.5 %
Loss on drying ≤0.5 % (105 °C, 3 hrs)
Sulfated ash ≤0.1 % (800 °C)
Specific optical rotation -15.5° to -17.5° (20 °C, 589 nm) (c=4, H2O)
Arsenic (As) ≤2 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR