Eugenol 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 4-Allyl-2-methoxyphenol, 5-Allyl-2-hydroxyanisole

119110050 119111000 119115000 119110025
ACRO119110050EA 22.9 GBP
ACRO119110050 ACRO119111000 ACRO119115000 ACRO119110025
Eugenol 99%
Eugenol
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 254 °C (760 mmHg)
Melting Pt: 12…10 °C
Density: 1.06 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00008654
CAS Number: 97-53-0
EINECS: 202-589-1
Merck Index: 13,03931

Order Now

Specification Test Results

Appearance Clear colorless to yellow-green Liquid
Infrared spectrum Conforms
Refractive index 1.5400 to 1.5420 (20°C, 589 nm)
GC ≥98.5 %
Specific gravity (20°C) 1.064 to 1.070


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR