(Ethyl)triphenylphosphonium bromide 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Ethyltriphenylphosphonium bromide

119040250 119041000 119045000
ACRO119040250EA 23 GBP
ACRO119040250 ACRO119041000 ACRO119045000
(Ethyl)triphenylphosphonium bromide 98%
(Ethyl)triphenylphosphonium bromide
Formula: C₂₀H₂₀BrP
MW: 371.26 g/mol
Melting Pt: 206.5…208.5 °C
Flash Pt: >200 °C
Storage Temperature: Ambient
MDL Number: MFCD00011838
CAS Number: 1530-32-1
EINECS: 216-223-3
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 206.5 to 208.5 °C
Titration Argentometric ≥97.5 % (on dry substance)
Loss on drying ≤1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR