Ethyl-4-piperidinecarboxylate 98+%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Ethyl piperidine-4-carboxylate, Ethyl isonipecotate, Ethyl 4-Piperidinecarboxylate

118640250 118641000
ACRO118640250EA 28.7 GBP
ACRO118640250 ACRO118641000
Ethyl-4-piperidinecarboxylate 98+%
Ethyl 4-piperidinecarboxylate
Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
Boiling Pt: 95…98 °C (7 mmHg)
Density: 1.02 g/cm³
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00006003
CAS Number: 1126-09-6
EINECS: 214-416-7

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4585 to 1.4605 (20°C, 589 nm)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR