2,6-Xylenol 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

116570010 116570050 116572500 116570025 116570051
ACRO116570010EA 57.5 GBP
ACRO116570010 ACRO116570050 ACRO116572500 ACRO116570025 ACRO116570051
2,6-Xylenol 99%
2,6-Xylenol
Formula: C₈H₁₀O
Boiling Pt: 203 °C (760 mmHg)
Melting Pt: 45…48 °C
Density: 1.15 g/cm³
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00002240
CAS Number: 576-26-1
EINECS: 209-400-1
UN: 2261
ADR: 6.1,II
Merck Index: 12,10215

Order Now

Specification Test Results

Appearance Colorless to yellow-brown Low melting solid (May darken on storage)
Infrared spectrum Conforms
Melting point 45 to 48 °C
Water ≤0.05 %
GC ≥98.5 %
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤3 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %
Iodine consuming subst. ≤0.004 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR