2-Chloroethyldimethylammonium chloride 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: N-(2-Chloroethyl)-N,N-Dimethylammonium Chloride, 2-Dimethylaminoethyl chloride hydrochloride

115820050 115821000 115825000 115820025 115820010
ACRO115820050EA 14.8 GBP
ACRO115820050 ACRO115821000 ACRO115825000 ACRO115820025 115820010.
2-Chloroethyldimethylammonium chloride 99%
2-Chloroethyldimethylammonium chloride
Formula: C₄H₁₁Cl₂N
MW: 144.04 g/mol
Melting Pt: 203…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00012516
CAS Number: 4584-46-7
EINECS: 224-970-1
UN: 3077
ADR: 9 III

Order Now

Specification Test Results

Appearance White to brown-yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 203 to 210 °C
Water ≤1.0 % (K.F.)
Titration with NaOH 98.5 to 101.5 % (On dry substance)
Chloride (Cl) 48.4 to 50.0 % (total chloride)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR