1,6-Diisocyanatohexane 99+%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

115190250 115191000 115190010
ACRO115190250EA 26.4 GBP
ACRO115190250 ACRO115191000 ACRO115190010
1,6-Diisocyanatohexane 99+%
1,6-Diisocyanatohexane
Formula: C₈H₁₂N₂O₂
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: –67 °C
Density: 1.04 g/cm³
Flash Pt: 130 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002047
CAS Number: 822-06-0
EINECS: 212-485-8
UN: 2281
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4520 to 1.4540 (20°C, 589 nm)
GC ≥99.0 %
Stabilizer ≤1 % (Type of stabilizer mentioned on the coa)
Residue after evaporation ≤0.001 %
Free halogens Passes test
Heavy metals (as Pb) ≤1 ppm
Titrable acid ≤0.0001 meq/g
Titrable base ≤0.0003 meq/g

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR