1,3-Dicyclohexylurea 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: N,N′-Dicyclohexylurea, N,N'-Dicyclohexylurea

113910250 113911000
ACRO113910250EA 42.6 GBP
ACRO113910250 ACRO113911000
1,3-Dicyclohexylurea 98%
1,3-Dicyclohexylurea
Formula: C₁₃H₂₄N₂O
Melting Pt: 231…235 °C
Storage Temperature: Ambient
MDL Number: MFCD00003829
CAS Number: 2387-23-7
EINECS: 219-213-7

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 231 to 236 °C
Titration with HClO4 ≥97.5 %
HPLC ≥99.0 % (ELSD)
Dnase,Rnase,Protease None detected
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR