3,4-Diaminobenzoic acid 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

112100250 112101000 112105000
ACRO112100250EA 26.8 GBP
ACRO112100250 ACRO112101000 ACRO112105000
3,4-Diaminobenzoic acid 97%
3,4-Diaminobenzoic acid
Formula: C₇H₈N₂O₂
Boiling Pt: 354 °C (1013 hPa)
Melting Pt: 215 °C (dec.)
Density: 1,544 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007726
CAS Number: 619-05-6
EINECS: 210-577-2

Order Now

Specification Test Results

Appearance Brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Titration with NaOH 96.0 to 104.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR