Cyclopentane 97%, extra pure

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Pentamethylene

111480010 111480025 111481000
ACRO111480010EA 136 GBP
ACRO111480010 ACRO111480025 ACRO111481000
Cyclopentane 97%, extra pure
Cyclopentane
Formula: C₅H₁₀
MW: 70.13 g/mol
Boiling Pt: 50 °C (760 mmHg)
Melting Pt: –94 °C
Density: 0.75 g/cm³
Flash Pt: –7 °C
Storage Temperature: Ambient
MDL Number: MFCD00001356
CAS Number: 287-92-3
EINECS: 206-016-6
UN: 1146
ADR: 3 II
Merck Index: 13,02769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4dd230057521cc77c90840c7adfece9c74c0a3b1&vid=352021851b7701a448c5b11d43e94d925d03f802&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4050 to 1.4070 (20°C, 589 nm)
GC ≥96.0 %
Residue after evaporation ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR