Citral cis- and trans-mixture 95%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

110440050 110441000 110445000
ACRO110440050EA 18.3 GBP
ACRO110440050 ACRO110441000 ACRO110445000
Citral cis- and trans-mixture 95%
Citral
Formula: C₁₀H₁₆O
Boiling Pt: 229 °C (1013 hPa)
Melting Pt: –10 °C
Density: 0.889 g/cm³
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00006997
CAS Number: 5392-40-5
EINECS: 226-394-6
Merck Index: 13,02346

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4860 to 1.4900 (20°C, 589 nm)
Density (20/4°C) 0.886 to 0.893 kg/l
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR