4-Chloro-3,5-xylenol 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

109061000 109065000 109060025
ACRO109061000EA 16.2 GBP
ACRO109061000 ACRO109065000 ACRO109060025
4-Chloro-3,5-xylenol 99%
4-Chloro-3,5-xylenol
Formula: C₈H₉ClO
Boiling Pt: 246 °C (760 mmHg)
Melting Pt: 113…116 °C
Density: 1,364 g/cm³ (20 °C)
Flash Pt: 132 °C
Storage Temperature: Ambient
MDL Number: MFCD00002324
CAS Number: 88-04-0
EINECS: 201-793-8
UN: 3077
ADR: 9,III
Merck Index: 13,02194

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 113 to 116 °C
GC ≥98.5 %
Stabilizer ca. 500 ppm MEHQ


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR