4-Chloroaniline 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

108590010 108590050 108591000 108595000
ACRO108590010EA 92.4 GBP
ACRO108590010 ACRO108590050 ACRO108591000 ACRO108595000
4-Chloroaniline 98%
4-Chloroaniline
Formula: C₆H₆ClN
Boiling Pt: 232 °C (760 mmHg)
Melting Pt: 68…72 °C
Density: 1,43 g/cm³ (20 °C)
Flash Pt: >188 °C
Storage Temperature: Ambient
MDL Number: MFCD00007835
CAS Number: 106-47-8
EINECS: 203-401-0
UN: 2018
ADR: 6.1,II
Merck Index: 13,02136

Order Now

Specification Test Results

Appearance Beige to brown to purple Crystalline solid (May darken on storage)
Infrared spectrum Conforms
Melting point 68 to 72 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR