(1S)-(+)-Camphor-10-sulphonic acid 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: ((1S,4R)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonic acid, (1S)-(+)-2-Oxobornane-10-sulphonic acid, D(+)-Camphor-10-sulphonic acid, (1S)-(+)-7,7-Dimethylbicyclo(2.2.1)-2-heptanon-1-methane sulphonic acid, (1S)-(+)-10-Camphorsulphonic acid, D-Camphorsulphonic acid

108221000 108225000
ACRO108221000EA 46.7 GBP
ACRO108221000 ACRO108225000
(1S)-(+)-Camphor-10-sulphonic acid 99%
(1S)-(+)-Camphor-10-sulphonic acid
Formula: C₁₀H₁₆O₄S
MW: 232.3 g/mol
Boiling Pt: 375
Melting Pt: >190 °C (dec.)
MDL Number: MFCD00064157
CAS Number: 3144-16-9
EINECS: 221-554-1
UN: 3261

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point ≥190 °C
Loss on drying ≤2 % (1 g, 105 °C, 5 hrs)
Specific optical rotation +21° to +23° (25 °C, 589 nm) (c=20, H2O)
Titration with NaOH 98.5 to 101.5 % (On dried substance)
Sulfate (SO4) ≤500 ppm
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR