4-tert-Butylphenol 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

108000010 108000025 108000050
ACRO108000010EA 30.9 GBP
ACRO108000010 ACRO108000025 ACRO108000050
4-tert-Butylphenol 97%
4-tert-Butylphenol
Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 237
Melting Pt: 96…100 °C
Density: 0.9 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002367
CAS Number: 98-54-4
EINECS: 202-679-0
UN: 3077
ADR: 9 III
Merck Index: 13,01584

Order Now

Specification Test Results

Appearance White to light beige Flakes or pastilles
Infrared spectrum Conforms
Melting point 96 to 100 °C
Water ≤0.5 % (K.F.)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR