2-tert-Butyl-p-cresol 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 1-tert-Butyl-2-hydroxy-5-methylbenzene, 2-tert-Butyl-4-methylphenol, 3-tert-Butyl-4-hydroxytoluene

107960050 107961000 107965000
ACRO107960050EA 14.7 GBP
ACRO107960050 ACRO107961000 ACRO107965000
2-tert-Butyl-p-cresol 99%
2-tert-Butyl-p-cresol
Formula: C₁₁H₁₆O
Boiling Pt: 244 °C (1013 hPa)
Melting Pt: 50…53 °C
Density: 0,961 g/cm³ (20 °C)
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00002381
CAS Number: 2409-55-4
EINECS: 219-314-6
UN: 2430
ADR: 8,III

Order Now

Specification Test Results

Appearance White to yellow to light beige Crystalline mass
Infrared spectrum Conforms
Melting point 50 to 53 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR