Diacetyl 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

107650050 107651000 107655000
ACRO107650050EA 20.4 GBP
ACRO107650050 ACRO107651000 ACRO107655000
Diacetyl 99%
Diacetyl
Formula: C₄H₆O₂
Boiling Pt: 190 °C (1013 hPa)
Melting Pt: 4…2 °C
Density: 0.985 g/cm³
Flash Pt: 7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008756
CAS Number: 431-03-8
EINECS: 207-069-8
UN: 2346
ADR: 3,II
Merck Index: 13,02985

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3910 to 1.3990 (20°C, 589 nm)
GC ≥98.5 %
Specific gravity (20°C) 0.98 to 0.99
Suitability (for Cu + Fe determination) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR