1,4-Butanediol 99+%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

107630010 107630025 107630100
ACRO107630010EA 29.1 GBP
ACRO107630010 ACRO107630025 ACRO107630100
1,4-Butanediol 99+%
1,4-Butanediol
Formula: C₄H₁₀O₂
Boiling Pt: 229.2 °C (1013hPa)
Melting Pt: 20 °C
Density: 1.01 g/cm³
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00002968
CAS Number: 110-63-4
EINECS: 203-786-5

Order Now

Specification Test Results

Appearance Clear viscous liquid or low melting solid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4470 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥99.0 %
Color scale ≤10 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR