4-Benzoylbutyric acid 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

105740250
ACRO105740250EA 96.3 GBP
ACRO105740250
4-Benzoylbutyric acid 97%
4-Benzoylbutyric acid

Appearance: Off-white to light beige Crystalline powder

Formula: C₁₁H₁₂O₃
Melting Pt: 124…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00004411
CAS Number: 1501-05-9

Order Now

Specification Test Results

Appearance Off-white to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 124 to 128 °C
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR