Benzophenone 99%, pure

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Diphenylmethanone, Diphenyl ketone

105560010 105565000 105560050
ACRO105560010EA 37.3 GBP
ACRO105560010 ACRO105565000 ACRO105560050
Benzophenone 99%, pure
Benzophenone
Formula: C₁₃H₁₀O
Boiling Pt: 305 °C (760 mmHg)
Melting Pt: 47…49 °C
Density: 1,085 g/cm³ (20 °C)
Flash Pt: 143 °C
Storage Temperature: Ambient
MDL Number: MFCD00003076
CAS Number: 119-61-9
EINECS: 204-337-6
ADR: 9,III
Merck Index: 13,01099

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 47 to 49 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR