Avantor® ACE® C18, HPLC Columns, 10 µm
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ACE-131-0346EA 555 GBP
ACE-131-0346 ACE-131-0510 ACE-131-0520 ACE-131-0530 ACE-131-1010 ACE-131-1020 ACE-131-1030 ACE-131-1046 ACE-131-1220 ACE-131-1246 ACE-131-1508 ACE-131-1510 ACE-131-1520 ACE-131-1530 ACE-131-1546 ACE-131-2510 ACE-131-2520 ACE-131-2530 ACE-131-2546 ACE-131-7520 ACE-131-0103GD ACE-131-0110GD ACE-131-7530 ACE-131-3020 ACE-131-3030 ACE-131-3046 ACE-131-3039 ACE-131-1004 ACE-131-1204 ACE-131-1503 ACE-131-1504 ACE-131-2046 ACE-131-2504 ACE-131-3004 ACE-131-0120GD
Avantor® ACE® C18, HPLC Columns, 10 µm
Chromatography Columns
Avantor® ACE® C18 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (3, 5 and 10 µm) and complementary phases
  • Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range ofMore Product Information