Anti-LUM Rabbit Recombinant Antibody [clone: R02-7F6]

Supplier: Abnova
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

RAB02125.100UL
ABNORAB02125.100ULEA 517 GBP
ABNORAB02125.100UL
Anti-LUM Rabbit Recombinant Antibody [clone: R02-7F6]
Antibodies
Order Now

SPECIFICATIONS

Antigen symbol LUM
Environmentally Preferable
Clonality Recombinant
Clone R02-7F6
Reactivity Human
Host Rabbit
Isotype
Immunogen Recombinant rabbit monoclonal antibody raised against human LUM.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR