Anti-17-alpha-Trenbolone Polyclonal Antibody

Supplier: Abnova
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PAB8204
ABNOPAB8204EA 1160 GBP
ABNOPAB8204
Anti-17-alpha-Trenbolone Polyclonal Antibody
Antibodies
Sheep polyclonal antibody raised against 17 alpha-Trenbolone.

Recommended Dilutions: Enzyme Immunoassay (0.5 ?g/ml

Type: Primary
Antigen: 17-alpha-Trenbolone
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR