Anti-PTPRC Mouse Monoclonal Antibody [clone: BRA-11]

Supplier: Abnova
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

MAB12142
ABNOMAB12142EA 517 GBP
ABNOMAB12142
Anti-PTPRC Mouse Monoclonal Antibody [clone: BRA-11]
Antibodies
Mouse monoclonal antibody raised against human PTPRC.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: BRA-11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol PTPRC
Antigen name Protein tyrosine phosphatase receptor type C
Environmentally Preferable
Antigen synonyms CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone BRA-11
Reactivity
Host Mouse
Gene ID 5788
Isotype IgG1 kappa
Western blot Yes
ImmunoChemistry Yes
Immunogen Non-T, non-B CALLA positive, ALL cell line REH.
Format Liquid
Purification Protein A/G purification
Cross adsorption No
Storage buffer PBS (0,05% BSA, 0,05% sodium azide).
Storage temperature Store at 4 °C. Aliquot to avoid repeated freezing and thawing.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR