Anti-PABPC5 (poly(A) binding Protein, cytoplasmic 5) Mouse Polyclonal Antibody

Supplier: Abnova
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H00140886-B01P
ABNOH00140886-B01PEA 393 GBP
ABNOH00140886-B01P
Anti-PABPC5 (poly(A) binding Protein, cytoplasmic 5) Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PABPC5 (poly(A) binding Protein, cytoplasmic 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR