Anti-PFKFB3 Rabbit Polyclonal Antibody

Supplier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AP8145B
ABGEAP8145BEA 345 GBP
ABGEAP8145B
Anti-PFKFB3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PFKFB3 (6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

SPECIFICATIONS

Antigen symbol PFKFB3
Environmentally Preferable
Antigen name 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3
Antigen synonyms 6PF-2-K/Fru-2,6-P2ASE brain/placenta-type isozyme|Fructose-2,6-bisphosphatase|iPFK-2|6-phosphofructo-2-kinase|IPFK2|PFK2|6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Mouse
Host Rabbit
Gene ID 6439
Isotype
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Epitope C-Terminal (450–550 aa)
Immunogen This SFTPB antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 129-158 amino acids from the Central region of human SFTPB.
Sequence KSRQPEPEQEPGMSDC
Purification Protein A & Peptide affinity purification
Concentration 0,5 mg/ml
Cross adsorption No
Storage buffer PBS with 0,09% (W/V) sodium azide, pH 7,4
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR